Learn More
Bellco Glass, Inc. Compression Fittng, 1/4in.
SDP
Catalog No. 501944921
Encompass_Preferred
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200mm stainless steal or a 45mm vertical sidearm. MADE IN THE USA. Specifications:316L Stainless SteelSize: 1/4"
Catalog No. 50-194-4921 Supplier Bellco Glass, Inc. Supplier No. 196490015
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200mm stainless steal or a 45mm vertical sidearm. MADE IN THE USA. Specifications:316L Stainless SteelSize: 1/4"