Learn More
Chemglass Life Sciences 1/2" PFA Compression Nut, Ferrule and Grip Ring
SDP
Catalog No. 50143647
Encompass_Preferred
1/2" PFA Compression Nut, Ferrule and Grip Ring. PFA Compression nut, PTFE ferrule and ETFE gripping ring for CG-1975 series compression fittings.
Catalog No. 50-143-647 Supplier Chemglass Life Sciences Supplier No. CG1975N20
1/2" PFA Compression Nut, Ferrule and Grip Ring. PFA Compression nut, PTFE ferrule and ETFE gripping ring for CG-1975 series compression fittings.