Learn More
Chemglass Life Sciences 3/4" PFA Compression Nut, Ferrule and Grip Ring
SDP
Catalog No. 50143653
Encompass_Preferred
3/4" PFA Compression Nut, Ferrule and Grip Ring. PFA Compression nut, PTFE ferrule and ETFE gripping ring for CG-1975 series compression fittings.
Catalog No. 50-143-653 Supplier Chemglass Life Sciences Supplier No. CG1975N22
3/4" PFA Compression Nut, Ferrule and Grip Ring. PFA Compression nut, PTFE ferrule and ETFE gripping ring for CG-1975 series compression fittings.