Learn More
Med Vet International Compression Plate, 2.0mm, 7 holes, 37mm, J0417D, Each
Catalog No. 5000582814
Encompass_Preferred
2mm Compression Plate. Use with 2mm cortical screws ifcompression is required.
Catalog No. 50-005-82814 Supplier Med Vet International Supplier No. JOR417D
2mm Compression Plate. Use with 2mm cortical screws ifcompression is required.