Learn More
Med Vet International Compression Plate 3.5mm, 2 holes, 26mm L, J0422A
Catalog No. 5000581939
Encompass_Preferred
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.
Catalog No. 50-005-81939 Supplier Med Vet International Supplier No. JOR422A
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.