Learn More
Med Vet International Compression Plate 3.5mm, 8 holes, 98mm, J0422G
Catalog No. 5000582831
Encompass_Preferred
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.
Catalog No. 50-005-82831 Supplier Med Vet International Supplier No. JOR422G
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.