Learn More
Med Vet International Compression Plate 4.5mm, 8 holes, 135mm L, J0427F
Catalog No. 5000582380
Encompass_Preferred
Use with 4.5mm cortical screws to produce fracture-site compression.
Catalog No. 50-005-82380 Supplier Med Vet International Supplier No. JOR427F
Use with 4.5mm cortical screws to produce fracture-site compression.