Learn More
Med Vet International Compression Plate, J0420C, 5 Holes, 2.7mm x 44mm, Each
Catalog No. 5000582725
Encompass_Preferred
Use with 2.7mm cortical screws to produce fracture-site compression.
Catalog No. 50-005-82725 Supplier Med Vet International Supplier No. JOR420C
Use with 2.7mm cortical screws to produce fracture-site compression.