Learn More
Med Vet International Compression Plate, J0420F, 8 Holes, 2.7mm x 68mm, Each
Catalog No. 5000582848
Encompass_Preferred
Use with 2.7mm cortical screws to produce fracture-site compression.
Catalog No. 50-005-82848 Supplier Med Vet International Supplier No. JOR420F
Use with 2.7mm cortical screws to produce fracture-site compression.