Learn More
Med Vet International Compression Plate, J0420H, 10 Holes, 2.7mm x 84mm, Each
Catalog No. 5000582258
Encompass_Preferred
Use with 2.7mm cortical screws to produce fracture-site compression.
Catalog No. 50-005-82258 Supplier Med Vet International Supplier No. JOR420H
Use with 2.7mm cortical screws to produce fracture-site compression.