Learn More
Med Vet International Compression Plate, J0420I, 12 Holes, 2.7mm x 100mm, Each
Catalog No. 5000581938
Encompass_Preferred
Use with 2.7mm cortical screws to produce fracture-site compression.
Catalog No. 50-005-81938 Supplier Med Vet International Supplier No. JOR420I
Use with 2.7mm cortical screws to produce fracture-site compression.