Learn More
Waters Corp Waters Corporation Compression Screw, 1/8 in, PEEK, Fitting
Catalog No. 50793836 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This compression screw is a 1/4-28 flangeless compression screw with an inner diameter of 1/8 " It is used for locking style degasser fittings.
  • Made from PEEK material.
  • Designed as a fitting.
  • Compatible with HPLC and UPLC systems.
  • Supplied as 1 unit per package.
  • Features a 1/4-28 flangeless design.
  • Used for locking style degasser fittings.
Catalog No. 50-793-836 Supplier Waters Corp Supplier No. WAT271028
This compression screw is a 1/4-28 flangeless compression screw with an inner diameter of 1/8 " It is used for locking style degasser fittings.
  • Made from PEEK material.
  • Designed as a fitting.
  • Compatible with HPLC and UPLC systems.
  • Supplied as 1 unit per package.
  • Features a 1/4-28 flangeless design.
  • Used for locking style degasser fittings.