Learn More
Waters Corp Waters Corporation Compression Screw and Ferrule, PEEK, With Lock Ring, 5 pk
Catalog No. 50792496 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation PEEK Compression Screw and Ferrule is an essential fitting designed for secure connections in various liquid chromatography systems. This product is provided in a package of 5 units, each featuring a lock ring for enhanced stability and reliability.
  • Made of PEEK material for chemical compatibility and durability.
  • Includes a lock ring for a secure fit.
  • Supplied as 5 units per package.
  • Compatible with UPLC components such as ASM, H-Class QSM, QSM, and TUV Detector.
  • Suitable for HPLC, MS, purification, and UPLC system types.
  • Supports a broad range of liquid chromatography instruments.
  • Designed for replacement as required.
Catalog No. 50-792-496 Supplier Waters Corp Supplier No. 700000991
The Waters Corporation PEEK Compression Screw and Ferrule is an essential fitting designed for secure connections in various liquid chromatography systems. This product is provided in a package of 5 units, each featuring a lock ring for enhanced stability and reliability.
  • Made of PEEK material for chemical compatibility and durability.
  • Includes a lock ring for a secure fit.
  • Supplied as 5 units per package.
  • Compatible with UPLC components such as ASM, H-Class QSM, QSM, and TUV Detector.
  • Suitable for HPLC, MS, purification, and UPLC system types.
  • Supports a broad range of liquid chromatography instruments.
  • Designed for replacement as required.