Learn More
Med Vet International 2 Layer Compression Bandage System 3M Coban 2 4 inch X 3-4/5 Yard / 4 inch X 6-3/10 Yard Self-Adherent / Pull On Closure Tan / White Nonsterile 35 To 40 mmhg, 2094Xl, 8 Kits/Case
Catalog No. 502830597
Encompass_Preferred
This Coban™ 2-Layer compression bandage delivers sustained therapeutic compression of 35 to 40 mmHg for up to 7 days It consists of a 4-inch × 3-4/5-yard comfort layer and a 4-inch × 6-3/10-yard compression layer for better results and wearability This product can be fully stretched over an affected area of skin This tan bandage is lightweight and breathable enough for wearing with regular shoes It is free of natural rubber for greater comfort
Catalog No. 50-283-0597 Supplier Med Vet International Supplier No. 993482CS
This Coban™ 2-Layer compression bandage delivers sustained therapeutic compression of 35 to 40 mmHg for up to 7 days It consists of a 4-inch × 3-4/5-yard comfort layer and a 4-inch × 6-3/10-yard compression layer for better results and wearability This product can be fully stretched over an affected area of skin This tan bandage is lightweight and breathable enough for wearing with regular shoes It is free of natural rubber for greater comfort