Learn More
Med Vet International 2 Layer Compression Bandage System 3M Coban2 Lite 4 inch X 2-9/10 Yard / 4 inch X 5-1/10 Yard Self-Adherent, White, 25 To 30 mmhg, 2794N, 8 Kits/Case
Catalog No. 502830641
Encompass_Preferred
This Coban™ 2-Layer compression bandage delivers sustained therapeutic compression of 25 to 30 mmHg for up to 7 days It consists of a 4-inch × 2-9/10-yard comfort layer and a 4-inch × 5-1/10-yard compression layer for better results and wearability This product can be fully stretched over an affected area of skin This tan and white compression bandage is lightweight and breathable enough for wearing with regular shoes It is free of natural rubber for greater comfort First layer is white and second layer is tan
Catalog No. 50-283-0641 Supplier Med Vet International Supplier No. 989622CS
This Coban™ 2-Layer compression bandage delivers sustained therapeutic compression of 25 to 30 mmHg for up to 7 days It consists of a 4-inch × 2-9/10-yard comfort layer and a 4-inch × 5-1/10-yard compression layer for better results and wearability This product can be fully stretched over an affected area of skin This tan and white compression bandage is lightweight and breathable enough for wearing with regular shoes It is free of natural rubber for greater comfort First layer is white and second layer is tan