Learn More
Bellco Glass, Inc. Compression Fitting,1/2in. Large Flange - Repl Part
SDP
Catalog No. 501944935
Encompass_Preferred
Large flange headplate assemblies.Universal headplates fit 5l thru 36l vessels. MADE IN THE USA. Corrosion resistant 316l stainless steel.These thermowells are designed specifically for the Bellco control systems and fit through a 5/16" fitting on the stainless steel headplate. See the BCS section for complete listings. Probe adapters & compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm.
Catalog No. 50-194-4935 Supplier Bellco Glass, Inc. Supplier No. 196490120
Large flange headplate assemblies.Universal headplates fit 5l thru 36l vessels. MADE IN THE USA. Corrosion resistant 316l stainless steel.These thermowells are designed specifically for the Bellco control systems and fit through a 5/16" fitting on the stainless steel headplate. See the BCS section for complete listings. Probe adapters & compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm.