Learn More
Bellco Glass, Inc. Compression Fitting 2.5"
SDP
Catalog No. 501944950
Encompass_Preferred
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. 1/2" pipe thread screws directly into headplate. MADE IN THE USA.
Catalog No. 50-194-4950 Supplier Bellco Glass, Inc. Supplier No. 196492000
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. 1/2" pipe thread screws directly into headplate. MADE IN THE USA.