Learn More
Grainger Compressor Vacuum Pump, 1/8 HP
Catalog No. NC2207428
Encompass_Preferred
The Vacuum Pump is a 1/8 HP oil-less diaphragm vacuum pump. It has a free air displacement of 4.9 CFM and a maximum vacuum of 25 inches of mercury. This pump can be used for vacuum packaging, vacuum clamping, and other vacuum applications. It has a 1/8 HP, 115V AC motor with thermal overload protection. The pump is constructed of aluminum and engineered plastics and has an on/off rocker switch. It measures 9 x 6 x 11 inches and weighs 13 pounds. The pump comes with a 2 year warranty.
This item is not returnable. View return policy
Catalog No. NC2207428 Supplier Grainger Supplier No. 3KYY6
The Vacuum Pump is a 1/8 HP oil-less diaphragm vacuum pump. It has a free air displacement of 4.9 CFM and a maximum vacuum of 25 inches of mercury. This pump can be used for vacuum packaging, vacuum clamping, and other vacuum applications. It has a 1/8 HP, 115V AC motor with thermal overload protection. The pump is constructed of aluminum and engineered plastics and has an on/off rocker switch. It measures 9 x 6 x 11 inches and weighs 13 pounds. The pump comes with a 2 year warranty.