Learn More
Precisionary Instruments Inc COMPRESSTOME SPEC TUBE 1X20MM
SDP
Catalog No. NC1941218
Encompass
Compresstome 310/210 Specimen Tube (20 mm diameter)Compresstome specimen tube with 20 mm diameter. Compatible with VF-210-0Z and VF-310-0Z onlyIncludes: 1 Specimen Tube (20mm) with white plunger / 5 Plastic Cleanup Razors
Catalog No. NC1941218 Supplier Precisionary Instruments Inc Supplier No. VFSPSVM20BOS
Compresstome 310/210 Specimen Tube (20 mm diameter)Compresstome specimen tube with 20 mm diameter. Compatible with VF-210-0Z and VF-310-0Z onlyIncludes: 1 Specimen Tube (20mm) with white plunger / 5 Plastic Cleanup Razors