Learn More
Medchemexpress LLC HY-P1036 500ug , Compstatin CAS:206645-99-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1036500UG
Encompass_Preferred
HY-P1036 500ug Compstatin CAS: 206645-99-0 Compstatin, a 13-residue cyclic peptide, is a potent inhibitor of the complement system C3 with species specificity. Compstatin binds to baboon C3 and is resistant to proteolytic cleavage in baboon blood (similar to humans). Compstatin inhibits only the activation of primates’ complement system. Compstatin exhibits IC50 values of 63 μM and 12 μM for classical and alterative complement pathway, respectively[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1984