Learn More
TARGETMOL CHEMICALS INC COMPSTATIN 5MG
SDP
Catalog No. 502578858
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Compstatin a cyclic tridecapeptide which was originally discovered from phage-display libraries is a highly potent and selective C3 inhibitor. purity: 100%
Catalog No. 50-257-8858 Supplier TARGETMOL CHEMICALS INC Supplier No. TP13045MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Compstatin a cyclic tridecapeptide which was originally discovered from phage-display libraries is a highly potent and selective C3 inhibitor. purity: 100%