Learn More
Selleck Chemical LLC Compstatin 5mg 206645-99-0
SDP
Supplier:  Selleck Chemical LLC S85225MG
Encompass_Preferred
Compstatin binds to complement component C3 and inhibits complement activation with IC50 of 12 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6101