Learn More
Bachem Compstatin Trifluoroacetate
SDP
Supplier:  Bachem 4045832.1000
Encompass_Preferred
Compstatin Trifluoroacetate, H-Ile-Cys-Val-Val-Gln-Asp-Trp-Gly-His-His-Arg-Cys-Thr-NH2, 206645-99-0, C66H99N23O17S2, Mr 1550.79, 1mg. Compstatin, ICVVQDWGHHRCT, is a 13-residue cyclic peptide that inhibits complement activation by binding to complement component C3.
Catalog No. 50-194-7449