Learn More
Medchemexpress LLC HY-P1036A 1mg Medchemexpress, Compstatin (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1036A1MG
Encompass_Preferred
Medchemexpress, HY-P1036A 1mg Compstatin (TFA) CAS: Compstatin TFA, a 13-residue cyclic peptide, is a potent inhibitor of the complement system C3 with species specificity. Compstatin TFA binds to baboon C3 and is resistant to proteolytic cleavage in baboon blood (similar to humans). Compstatin TFA inhibits only the activation of primates’ complement system. Compstatin TFA exhibits IC50 values of 63 μM and 12 μM for classical and alterative complement pathway, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3035