Learn More
Enterprise Technology Solutions Compucessory Flash Drive, 32GB, No Password Protectn, Black/Silver
SDP
Supplier:  Enterprise Technology Solutions CCS91007
Encompass_Preferred
Memory Stick-compliant flash drive offers portable storage in a compact package with plug-and-play convenience. Write speed is up to 5MB per second with USB 2.0 interface. Read speed is up to 12MB per second. 32GB flash drive is compatible with USB 2.0 and 1.1 USB ports as well as Windows ME, 2000, XP, Vista 7, Linux 2.4 or later, Mac 9.0 or later. Flash drive does not have password protection or encryption protection.
Catalog No. 50-007-39847