Learn More
Grainger Computer Desk 46 x 30 x 28 In Charcoal
Catalog No. 502764401
Encompass_Preferred
The Grainger 16W996 is a Philips 16-watt T8 fluorescent light bulb that provides 1600 lumens of light output. This 4 foot long bulb has a medium bi-pin base and a color temperature of 4100K. It has an average rated life of 20,000 hours and meets EISA standards for energy efficiency. The Philips 16W996 T8 is a good choice for energy efficient fluorescent lighting in commercial, industrial and residential applications.
Catalog No. 50-276-4401 Supplier Grainger Supplier No. 16W996
The Grainger 16W996 is a Philips 16-watt T8 fluorescent light bulb that provides 1600 lumens of light output. This 4 foot long bulb has a medium bi-pin base and a color temperature of 4100K. It has an average rated life of 20,000 hours and meets EISA standards for energy efficiency. The Philips 16W996 T8 is a good choice for energy efficient fluorescent lighting in commercial, industrial and residential applications.