Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Comu™

Catalog No. 712191100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-219-1100G 100 g
71-219-125G 25 g
71-219-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-219-1100G Supplier Sigma Aldrich Supplier No. 712191100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a Safer, non-explosive and more effective replacement for Benzotriazole-Based peptide coupling reagents and thus aligns with Less Hazardous Chemical Synthesis and Inherently Safer Chemistry for Accident Prevention. Click here for more information.Morpholine-Based Immonium and Halogenoamidinium Salts as Coupling Reagents in Peptide SynthesisCOMU is a registered trademark of Luxembourg Bio Technologies Ltd.

Specifications

CAS 1075198-30-9
Molecular Formula C12H19F6N4O4P
Linear Formula C12H19F6N4O4P
MDL Number MFCD11975052
Quantity 100 g
Synonym (1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholino-carbenium hexafluorophosphate
Molecular Weight (g/mol) 428.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.