Learn More
Cole-Parmer Oakton CON 6+ Handheld Conductivity Meter with Probe
Supplier:  Cole-Parmer 3560400
Encompass_Preferred
These microprocessor-based meters feature easy, push-button operation that offers high accuracy at an extremely affordable price. A quick and easy single-point calibration covers all five ranges with the CON 6+ meters; for greater precision, single points can be calibrated within each of the five measurement ranges. Additional features include a convenient Hold function and auto-off function that shuts meter off after 20 minutes of nonuse. Programming functions allow customization of meter for temperature compensation, temperature coefficient, normalization, and cell constant for better accuracy in varying applications.
Catalog No. NC0346700