Learn More
Crystal Chem Inc Control for the Rat Total Bile Acid Kit
SDP
Catalog No. 501947711
Encompass_Preferred
The Control for the Rat Total Bile Acid Assay can be used to validate the calibration curve obtained when running the Rat Total Bile Acid Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7711 Supplier Crystal Chem Inc Supplier No. 80463
The Control for the Rat Total Bile Acid Assay can be used to validate the calibration curve obtained when running the Rat Total Bile Acid Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.