Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Con-Space™ SCBA Accessory, Facemask with ComSet/Speaker

Catalog No. 19808756
Change view
Click to view available options
With ComSet/Speaker
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-808-756 With ComSet/Speaker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-808-756 Supplier Con-Space™ Supplier No. 010304031

missing translation for 'validMainframeMsg'

Specifications

Color Black
Product Type Full Facepiece Respirator
For Use With (Application) Medical and security
For Use With (Equipment) SCBA
Includes Speaker, alarm button
Material Silicone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.