Learn More
Eberbach Corp Con-Torque Tissue Homogenizer
SDP
Catalog No. NC9848092
Encompass_Preferred
Variable-speed Con-Torque Tissue Homogenizer gives 196 oz-in of continuous torque at all speeds providing continuous, effortless, positive grinding action. Full torque eliminates the possibility of tissue jamming the pestle in the grinding vessel. Totally enclosed, fan cooled 1/9 hp motor drives the Con-Torque power unit through a continuously variable transmission. Speeds from 0 to 400 rpm. Sintered bronze motor bearings require no lubrication. 3/8” output shaft provides rotation for the chuck through a flexible coupling allowing operation at various angles which reduces breakage of glassware. A knurled hand-wheel offers a counter support grip when tightening the chuck which accepts shafts up to 1/2” in diameter. 3/8” fixed coupling is included for techniques requiring application of side pressure to pestle and mortar. Integral clamp fits on support rods up to 3/4” in diameter. Includes Con-Torque power unit, chucking device and universal support stand, but without mortar or pestle.
Catalog No. NC9848092 Supplier Eberbach Corp Supplier No. E2355.00
Variable-speed Con-Torque Tissue Homogenizer gives 196 oz-in of continuous torque at all speeds providing continuous, effortless, positive grinding action. Full torque eliminates the possibility of tissue jamming the pestle in the grinding vessel. Totally enclosed, fan cooled 1/9 hp motor drives the Con-Torque power unit through a continuously variable transmission. Speeds from 0 to 400 rpm. Sintered bronze motor bearings require no lubrication. 3/8” output shaft provides rotation for the chuck through a flexible coupling allowing operation at various angles which reduces breakage of glassware. A knurled hand-wheel offers a counter support grip when tightening the chuck which accepts shafts up to 1/2” in diameter. 3/8” fixed coupling is included for techniques requiring application of side pressure to pestle and mortar. Integral clamp fits on support rods up to 3/4” in diameter. Includes Con-Torque power unit, chucking device and universal support stand, but without mortar or pestle.