Learn More
Cole-Parmer Oakton CON 700 Meter with Probe, Stand, and NIST-Traceable Calibration
Catalog No. 501961743 Shop All Cole-Parmer Products
Encompass_Preferred
Proviing an affordable way to take consistent, accurate conductivity measurements in your laboratory, the CON 700 meter reads 0 to 200 mS across five ranges, with an autoranging feature that automatically gives the best resolution for your measurements. Calibrate at up to five conductivity points (one per range) for improved measurement accuracy. Adjustable temperature coefficient provides additional accuracy. Cell constant is selectable for very high or low range measurements - meter is flexible to your application. Selectable manual or automatic temperature compensation gives greater precision. Meter is encased in a compact 6-1/8" L x 6-7/8" W x 2-3/4" (15.5 x 17.5 x 6.9 cm) housing which takes up a minimum of bench space-nearly 40% less space than comparable meters. Water-resistant membrane keypad and convenient pullout reference guide provide useful features for the lab.
Catalog No. 50-196-1743 Supplier Cole-Parmer Supplier No. 3541101
Proviing an affordable way to take consistent, accurate conductivity measurements in your laboratory, the CON 700 meter reads 0 to 200 mS across five ranges, with an autoranging feature that automatically gives the best resolution for your measurements. Calibrate at up to five conductivity points (one per range) for improved measurement accuracy. Adjustable temperature coefficient provides additional accuracy. Cell constant is selectable for very high or low range measurements - meter is flexible to your application. Selectable manual or automatic temperature compensation gives greater precision. Meter is encased in a compact 6-1/8" L x 6-7/8" W x 2-3/4" (15.5 x 17.5 x 6.9 cm) housing which takes up a minimum of bench space-nearly 40% less space than comparable meters. Water-resistant membrane keypad and convenient pullout reference guide provide useful features for the lab.