Learn More
MSC Ashburn Chemical Technologies All-Purpose Degreaser, Liquid Concentrate, 5 gal Pail, Lemon Scent
Catalog No. 502836958
Encompass
This all-purpose cleaner and degreaser is a biodegradable and non-toxic solution designed to effectively cut through, lift, and suspend grease, dirt, soil, and grime. Presented as an environmentally safe alternative to harsh solvents, this liquid concentrate features a pleasant lemon scent and is packaged in a convenient 5 gal pail. It is suitable for a wide range of applications, including the cleaning and degreasing of ferrous parts.
  • Biodegradable and non-toxic formula
  • Cuts through, lifts, and suspends grease, dirt, soil, and grime
  • Environmentally safe alternative to solvents and hazardous chemicals
  • Pleasant lemon scent
  • Provided as a liquid concentrate
  • Packaged in a 5 gal pail
  • Suitable for cleaning and degreasing ferrous parts
  • pH of 11.5
  • Completely soluble in water
Catalog No. 50-283-6958 Supplier MSC Supplier No. 65965287
This all-purpose cleaner and degreaser is a biodegradable and non-toxic solution designed to effectively cut through, lift, and suspend grease, dirt, soil, and grime. Presented as an environmentally safe alternative to harsh solvents, this liquid concentrate features a pleasant lemon scent and is packaged in a convenient 5 gal pail. It is suitable for a wide range of applications, including the cleaning and degreasing of ferrous parts.
  • Biodegradable and non-toxic formula
  • Cuts through, lifts, and suspends grease, dirt, soil, and grime
  • Environmentally safe alternative to solvents and hazardous chemicals
  • Pleasant lemon scent
  • Provided as a liquid concentrate
  • Packaged in a 5 gal pail
  • Suitable for cleaning and degreasing ferrous parts
  • pH of 11.5
  • Completely soluble in water