Learn More
Apexbio Technology LLC Concanavalin A (Con A)-FITC 5mg(5 mg/mL)
SDP
Catalog No. 503107012
Encompass_Preferred
Concanavalin A (Con A) derived from Canavalia ensiformis (jack bean) is a plant lectin protein with a molecular weight of approximately 104 kDa Each subunit can bind one Ca2 ion and one Mn2 ion and has one carbohydrate binding site Con A is capable of binding to various glycoproteins glycolipids and -D-glucose and -D-mannose residues FITC is a fluorescent dye that emits a green fluorescence under UV light (Ex/Em 495 nm/515 nm) This combination endows Con A-FITC with high visualization capability making it efficient for detecting carbohydrate molecules on the cell surface This product is FITC-labeled Con A in solution form It is applicable in glycobiology research serving as a probe to detect carbohydrate molecules on the cell surface and analyzed effectively using flow cytometry Applying IHC staining procedures with this product allows visualization of carbohydrate distribution in tissues or cells under a fluorescence microscope
Catalog No. 50-310-7012 Supplier Apexbio Technology LLC Supplier No. K441325
Concanavalin A (Con A) derived from Canavalia ensiformis (jack bean) is a plant lectin protein with a molecular weight of approximately 104 kDa Each subunit can bind one Ca2 ion and one Mn2 ion and has one carbohydrate binding site Con A is capable of binding to various glycoproteins glycolipids and -D-glucose and -D-mannose residues FITC is a fluorescent dye that emits a green fluorescence under UV light (Ex/Em 495 nm/515 nm) This combination endows Con A-FITC with high visualization capability making it efficient for detecting carbohydrate molecules on the cell surface This product is FITC-labeled Con A in solution form It is applicable in glycobiology research serving as a probe to detect carbohydrate molecules on the cell surface and analyzed effectively using flow cytometry Applying IHC staining procedures with this product allows visualization of carbohydrate distribution in tissues or cells under a fluorescence microscope