Learn More
Enzo Life Sciences Concanamycin A (25µg). CAS: 80890-47-7
SDP
Catalog No. 502008918
Encompass_Preferred
Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1 (BML-CM110). Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Exhibits cytotoxic effects in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production. Inhibits Autophagy. Alternative name: Folimycin. Purity: ≥98% (HPLC). Appearance: White to off-white solid (sub-milligram quantities will appear as clear film). Solubility: Soluble in methanol, DMSO or acetonitrile; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8918 Supplier Enzo Life Sciences Supplier No. ALX380034C025
Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1 (BML-CM110). Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Exhibits cytotoxic effects in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production. Inhibits Autophagy. Alternative name: Folimycin. Purity: ≥98% (HPLC). Appearance: White to off-white solid (sub-milligram quantities will appear as clear film). Solubility: Soluble in methanol, DMSO or acetonitrile; insoluble in water. Long Term Storage: -20°C.