Learn More
AdipoGen Concanamycin A (high purity),Chemical. CAS: 80890-47-7
SDP
Catalog No. 50464888
Encompass_Preferred
Concanamycin A (high purity), AdipoGen Life Sciences. Chemical. CAS: 80890-47-7. Formula: C46H75NO14. MW: 866.1. Isolated from Streptomyces sp. Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1 (Prod. No. BVT-0252). Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Cytotoxic in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production.
Catalog No. 50-464-888 Supplier AdipoGen Supplier No. BVT0237C025
Concanamycin A (high purity), AdipoGen Life Sciences. Chemical. CAS: 80890-47-7. Formula: C46H75NO14. MW: 866.1. Isolated from Streptomyces sp. Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1 (Prod. No. BVT-0252). Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Cytotoxic in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production.