Learn More
AdipoGen Concanamycin A high purity
SDP
Supplier:  AdipoGen BVT0237M005
Encompass_Preferred
Chemical. CAS 80890-47-7. Formula C46H75NO14. MW 866.1. Isolated from Streptomyces sp. Antibiotic. More potent and specific H-ATPase inhibitor than bafilomycin A1 Prod. No. BVT-0252. Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Cytotoxic in a number of cell lines in a cell viability assay. Induces nitric oxide NO production.
Catalog No. 50-256-2051