Learn More
AdipoGen Concanamycin B high purity
SDP
Catalog No. 502562042
Encompass_Preferred
Chemical. CAS 81552-33-2. Formula C45H73NO14. MW 852.1. Isolated from Streptomyces sp. Antibiotic. Exhibits similar activity as concanamycin A Prod. No. BVT-0237 and concanamycin C Prod. No. BVT-0254. Specific vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Suppresses bone resorption. Inhibits proliferation of mouse splenic lymphocytes. Antifungal and larvicidal.
Catalog No. 50-256-2042 Supplier AdipoGen Supplier No. BVT0253C500
Chemical. CAS 81552-33-2. Formula C45H73NO14. MW 852.1. Isolated from Streptomyces sp. Antibiotic. Exhibits similar activity as concanamycin A Prod. No. BVT-0237 and concanamycin C Prod. No. BVT-0254. Specific vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Suppresses bone resorption. Inhibits proliferation of mouse splenic lymphocytes. Antifungal and larvicidal.