Learn More
AdipoGen Concanamycin B high purity
SDP
Supplier:  AdipoGen BVT0253M001
Encompass_Preferred
Chemical. CAS 81552-33-2. Formula C45H73NO14. MW 852.1. Isolated from Streptomyces sp. Antibiotic. Exhibits similar activity as concanamycin A Prod. No. BVT-0237 and concanamycin C Prod. No. BVT-0254. Specific vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Suppresses bone resorption. Inhibits proliferation of mouse splenic lymphocytes. Antifungal and larvicidal.
Catalog No. 50-256-2041