Learn More
Cayman Chemical Concanavalin A, 11028-71-0, 100 mg
SDP
Supplier:  Cayman Chemical 14951100
Encompass_Preferred
A plant lectin that selectively cross-links cell-surface glycoproteins and affects the initiation of cell agglutination, mitogenesis, and apoptosis; widely used in affinity chromatography purifications of various glycoproteins and cellular structures; also used to induce liver injury in experimental mouse models of autoimmune hepatitis.
Catalog No. NC0968272