Learn More
Sigma Aldrich Fine Chemicals Biosciences Concanavalin A from Canavalia ensiformis (Jack bean), 11028-71-0, MFCD00071069, 5 mg
Catalog No. NC1491746
Encompass
Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture, Synonym: Con A. Concanavalin A from Canavalia ensiformis (Jack bean) has been used: in in vitro cell proliferation assay and cell cycle assessment, in lymphocyte proliferation, and in splenocyte culturing.
Catalog No. NC1491746 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C52755MG
Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture, Synonym: Con A. Concanavalin A from Canavalia ensiformis (Jack bean) has been used: in in vitro cell proliferation assay and cell cycle assessment, in lymphocyte proliferation, and in splenocyte culturing.