Learn More
Medchemexpress LLC Concanavalin A-AF488 | 1 MG
SDP
Catalog No. 5000425299
Encompass_Preferred
Concanavalin A-AF488 (Con A-AF488) is an AF488-labeled Concanavalin A. It functions as a Ca2+/Mn2+-dependent and mannose/glucose-binding plant lectin. This product is for research use only.
  • AF488-labeled lectin
  • Ca2+/Mn2+-dependent
  • Mannose/glucose-binding plant lectin
  • Binds N- and O-glycosylated proteins of head and neck carcinoma cells
Catalog No. 50-004-25299 Supplier Medchemexpress LLC Supplier No. HYNP164B1MG
Concanavalin A-AF488 (Con A-AF488) is an AF488-labeled Concanavalin A. It functions as a Ca2+/Mn2+-dependent and mannose/glucose-binding plant lectin. This product is for research use only.
  • AF488-labeled lectin
  • Ca2+/Mn2+-dependent
  • Mannose/glucose-binding plant lectin
  • Binds N- and O-glycosylated proteins of head and neck carcinoma cells