Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Concanavalin A from Canavalia Ensiformis Seeds (Jack Beans), MP Biomedicals

Catalog No. mp021507101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021507101 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021507101 Supplier MP Biomedicals, Inc Supplier No. 02150710.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Lectin are used in a wide variety of applications in vitro, including: blood grouping and erythrocyte polyagglutination studies, Mitogenic stimulation of lymphocytes, Lymphocyte subpopulation studies, Fractionation of cells and other particles, Histochemical studies of normal and pathological conditions
  • Con-A has been used to elucidate structural changes in the membrane surface of transformed cells
  • Con-A conjugates of antitumor drugs have been used in drug delivery systems in cultured cells

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mg
Product Type CONCANAVALIN A from Canavalia ensiformis seeds (jack beans)
Content And Storage -20°C
Form Powder
Color White
Solubility Soluble in water (10 mg/ mL-slightly hazy, colorless solution).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.