Learn More
Biotium Concanavalin A, CF405M Conjugate
Catalog No. 501964461
Encompass_Preferred
This product is fluorescently-labeled concanavalin A (Con A). Con A is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues, found in the cell wall of yeast and fungi, and the cell membrane of mammalian cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M dye has excitation/emission at 408/452 nm.
Catalog No. 50-196-4461 Supplier Biotium Supplier No. 29074
This product is fluorescently-labeled concanavalin A (Con A). Con A is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues, found in the cell wall of yeast and fungi, and the cell membrane of mammalian cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M dye has excitation/emission at 408/452 nm.