Learn More
Biotium Concanavalin A, CF405S Conjugate
Catalog No. 501964462
Encompass_Preferred
This product is fluorescently-labeled concanavalin A (Con A). Con A is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues, found in the cell wall of yeast and fungi, and the cell membrane of mammalian cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405S dye has excitation/emission at 404/431 nm.
Catalog No. 50-196-4462 Supplier Biotium Supplier No. 29075
This product is fluorescently-labeled concanavalin A (Con A). Con A is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues, found in the cell wall of yeast and fungi, and the cell membrane of mammalian cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405S dye has excitation/emission at 404/431 nm.