Learn More
Bioworld Concanavalin A (Jackbean) Lectin (Con A) - MicroPlate(s)s
Catalog No. 50197528
Encompass
High quality Con A coated microplates. Concanavalin A coated surfaces provide for effective and specific binding to the carbohydrate fraction of glycoproteins, enzymes and cell membranes.Technical Specifications:Ligand Source:Canavalia ensiformisMolecular Weight: 104 kDaCarbohydrate Specificity: αManInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AForm: 96 Well MicroplateIsoelectric (pI): 6.3-7Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-197-528 Supplier Bioworld Supplier No. 201400002
High quality Con A coated microplates. Concanavalin A coated surfaces provide for effective and specific binding to the carbohydrate fraction of glycoproteins, enzymes and cell membranes.Technical Specifications:Ligand Source:Canavalia ensiformisMolecular Weight: 104 kDaCarbohydrate Specificity: αManInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AForm: 96 Well MicroplateIsoelectric (pI): 6.3-7Storage: -20°CApplication note: Calcium and Manganese are required for binding.