Learn More
Biomatik Corporation Concanavalin A (ConA) ELISA Kit
Supplier:  Biomatik Corporation EKU03435
Encompass_Preferred
Concanavalin A (ConA) ELISA Kit for detecting ConA in biological fluids. Species Reactivity: Universal. Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Con-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9037