Learn More
Biomatik Corporation Concanavalin A (Cona) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU31541
Encompass_Preferred
Concanavalin A (Cona) Polyclonal Antibody is a Universal Polyclonal Antibody to Concanavalin A (ConA). The Concanavalin A (Cona) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Universal. Applications: WB, IHC, ICC, IP. Target Synonyms: Concanavalin A (ConA). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9204