Learn More
Sigma Aldrich Fine Chemicals Biosciences Concanavalin A from Canava1G
Catalog No. 502903413
Encompass
Concanavalin Anbsp(con A) is a plant lectin refined from jack beans. It is a tetrameric protein which has four identical 28 kDa subunits. Con A has the ability to connect with the mannose residues of several glycoproteins and can stimulate lymphocytes.
Catalog No. 50-290-3413 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C20101G
Concanavalin Anbsp(con A) is a plant lectin refined from jack beans. It is a tetrameric protein which has four identical 28 kDa subunits. Con A has the ability to connect with the mannose residues of several glycoproteins and can stimulate lymphocytes.