Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Concanavalin A, MP Biomedicals™

Catalog No. MP021507105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021507105 500 mg
MP021507102 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021507105 Supplier MP Biomedicals, Inc Supplier No. 02150710.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isolated by affinity chromatography on cross-linked dextran. Concanavalin A, MP Biomedicals is pure Canavalia ensiformis lectin (Con A) from Jackbean. It exists as a dimer below pH 5.0, and at pH >7, it exists as a tetramer.

Lectin are used in a wide variety of applications. Con-A is not a glycoprotein and does not contain cysteine residues. Unlike most other lectins, Con-A is a metalloprotein and requires a transition metal ion, such as manganese, plus calcium ions for binding.

  • Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Con-A binds specifically to mannosyl and glucosyl residues of polysaccharides and glycoproteins.
  • Unmodified hydroxyl groups at the C3, C4 and C6 positions of D-glucopyranosyl or D-mannopyranosyl rings may be essential for binding.
  • Each subunit of Con A contains one calcium ion and one manganese ion. Removal of these cations by dialysis under acidic conditions abolishes the carbohydrate-binding activity.
  • Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer. Above pH 7.0 higher aggregates are formed.
  • Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11028-71-0
Synonym Con A

Specifications

Quantity 500 mg
Solubility Information Soluble in water (10mg/mL, slightly hazy, colorless solution).
Formula Weight Monomeric molecular weight of Con-A is 25,500(Lit.)
Physical Form Powder
Recommended Storage −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.